missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Corticotropin-releasing factor receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Corticotropin-releasing factor receptor 1, was expressed in the hosts and tags indicated.
202.00€ - 2765.00€
Specifications
Accession Number | NP_001138618.1 |
---|---|
Gene ID (Entrez) | 104909134 |
Name | Corticotropin-releasing factor receptor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077581
|
enQuireBio™
QP9080-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16097581
|
enQuireBio™
QP9080-YE-1MG |
1 mg |
2765.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16087581
|
enQuireBio™
QP9080-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16007591
|
enQuireBio™
QP9080-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16017591
|
enQuireBio™
QP9080-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16027591
|
enQuireBio™
QP9080-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001138618.1 | |
Corticotropin-releasing factor receptor 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
104909134 | |
Research Use Only | |
CRHR1 | |
Recombinant Protein | |
Yeast | |
ASLQDQHCES LSLASNISGL QCNASVDLIG TCWPRSPAGQ LVVRPCPAFF YGVRYNTTNN GYRECLANGS WAARVNYSEC QEILNEEKKS KVHYHVAVI | |
Greater than 90% as determined by SDS-PAGE. |