missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COMM domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) COMM domain-containing protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8487-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024891

  • 530.00€ / 100µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001271306.1
COMM domain-containing protein 1
Research Use Only
COMMD4
Recombinant Protein
E. coli
MRFRFCGDLD CPDWVLAEIS TLAKMSSVKL RLLCSQVLKE LLGQGIDYEK ILKLTADAKF ESGDVKATVA VLSFILSSAA KHSVDGESLS SELQQLGLPK EHAASLCRCY EEKQSPLQKH LRVCSLRMNR LAGVGWRVDY TLSSSLLQSV EEPMVHLRLE VAAAPGTPAQ PVAMSLSADK FQVLLAELKQ AQTLM
Greater than 90% as determined by SDS-PAGE.
54939
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers