missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEBPG / CEBP gamma Protein

A DNA sequence encoding the Homo sapiens (Human) CEBPG / CEBP gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6946-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026820

  • 238.00€ / 10µg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001239225.1
CEBPG / CEBP gamma Protein
Research Use Only
CEBPG
Recombinant Protein
E. coli
MSKISQQNST PGVNGISVIH TQAHASGLQQ VPQLVPAGPG GGGKAVAPSK QSKKSSPMDR NSDEYRQRRE RNNMAVKKSR LKSKQKAQDT LQRVNQLKEE NERLEAKIKL LTKELSVLKD LFLEHAHNLA DNVQSISTEN TTADGDNA
Greater than 90% as determined by SDS-PAGE.
1054
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers