missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEBPG / CEBP gamma Protein

A DNA sequence encoding the Homo sapiens (Human) CEBPG / CEBP gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6946-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016820

Quantity Price
1 546.01€ / 100µg
6 513.42€ / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001239225.1
CEBPG / CEBP gamma Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1054
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CEBPG
Recombinant Protein
E. coli
MSKISQQNST PGVNGISVIH TQAHASGLQQ VPQLVPAGPG GGGKAVAPSK QSKKSSPMDR NSDEYRQRRE RNNMAVKKSR LKSKQKAQDT LQRVNQLKEE NERLEAKIKL LTKELSVLKD LFLEHAHNLA DNVQSISTEN TTADGDNA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers