missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD24 Fc Chimera Avi-tag Protein, CF
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human Siglec-10 Fc Chimera (Catalog # 2130-SL ) is coated at 5 μg/mL, the concentration of Biotinylated Recombinant Human CD24 Fc Chimera Avi-tag (Catalog # AVI5247) that produces 50% optimal binding response is found to be approximately 1.5-7.5 μg/mL.
Specifications
Specifications
| Accession Number | P25063.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 100133941 |
| Molecular Weight (g/mol) | M.W. (Observed): 43-55 kDa, under reducing conditions, M.W. (Theoretical): 32 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human CD24 protein Human CD24 (Ser27-Gly59) (N-terminus) Accession # P25063.3 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD 24, CD24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24A, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction