missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calponin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Calponin-2, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001290428.1 |
---|---|
Gene ID (Entrez) | 1265 |
Name | Calponin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002301
|
enQuireBio™
QP8036-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
16022301
|
enQuireBio™
QP8036-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16012301
|
enQuireBio™
QP8036-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16032301
|
enQuireBio™
QP8036-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042301
|
enQuireBio™
QP8036-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052301
|
enQuireBio™
QP8036-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001290428.1 | |
Calponin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1265 | |
Research Use Only | |
CNN2 | |
Recombinant Protein | |
E. coli | |
SSTQFNKGPS YGLSAEVKNR LLSKYDPQKE AELRTWIEGL TGLSIGPDFQ KGLKDGTILC TLMNKLQPGS VPKINRSMQN WHQLENLSNF IKAMVSYGMN PVDLFEANDL FESGNMTQVQ VSLLALAGKA KTKGLQSGVD IGVKYSEKQE RNFDDATMKA GQCVIGLQMG TNKCASQSGM TAYGTRRHLY DPKNHILPPM DHSTISLQMG TNKCASQVGM TAPGTRRHIY DTKLGTDKCD NSSMSLQMGY TQGANQSGQV FGLGRQIYDP KYCPQGTVAD GAPSGTGDCP DPGEVPEYPP YYQEEAGY | |
Greater than 90% as determined by SDS-PAGE. |