missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BEN domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) BEN domain-containing protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7716-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001073919.1 | |
BEN domain-containing protein 1 | |
Research Use Only | |
BEND3 | |
Recombinant Protein | |
Yeast | |
CLPQLNDFFS RFWAQREMED SQPSGQVASF FEAEQVDPGH FLDNKDQEEA LSLDRSSTIA SDHVVDTQDL TEFLDEASSP GEFAVFLLHR LFPELFDHRK LGEQYSCYGD GGKQELDPQR LQIIRNYTEI YFPD | |
Greater than 90% as determined by SDS-PAGE. |
57673 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |