missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl-2-like protein 15 Protein

A DNA sequence encoding the Homo sapiens (Human) Bcl-2-like protein 15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7698-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020271

  • 3405.00€ / 1mg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001010922.1
Bcl-2-like protein 15 Protein
Research Use Only
BCL2L15
Recombinant Protein
E. coli
MKSSQTFEEQ TECIVNTLLM DFLSPTLQVA SRNLCCVDEV DSGEPCSFDV AIIAGRLRML GDQFNGELEA SAKNVIAETI KGQTGAILQN TVESLSKTWC AQDSSLAYER AFLAVSVKLL EYMAHIAPEV VGQVAIPMTG MINGNQAIRE FIQGQGGWEN LES
Greater than 80% as determined by SDS-PAGE.
440603
1 mg
Not determined.
Full Length of BC127719
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers