missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Autophagy-related protein 16-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Autophagy-related protein 16-1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001177195.1 |
---|---|
Gene ID (Entrez) | 55054 |
Name | Autophagy-related protein 16-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030451
|
enQuireBio™
QP7727-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16050451
|
enQuireBio™
QP7727-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16040451
|
enQuireBio™
QP7727-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16060451
|
enQuireBio™
QP7727-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16070451
|
enQuireBio™
QP7727-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16080451
|
enQuireBio™
QP7727-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001177195.1 | |
Autophagy-related protein 16-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
55054 | |
Research Use Only | |
ATG16L1 | |
Recombinant Protein | |
E. coli | |
MSSGLRAADF PRWKRHISEQ LRRRDRLQRQ AFEEIILQYN KLLEKSDLHS VLAQKLQAEK HDVPNRHEIS PGHDGTWNDN QLQEMAQLRI KHQEELTELH KKRGELAQLV IDLNNQMQRK DREMQMNEAK IAECLQTISD LETECLDLRT KLCDLERANQ TLKDEYDALQ ITFTALEGKL RKTTEENQEL VTRWMAEKAQ EANRLNAENE KDSRRRQARL QKELAEAAKE PLPVEQDDDI EVIVDETSDH TEETSPVRAI SRAATKRLSQ PAGGLLDSIT NIFGRRSVSS FPVPQDNVDT HPGSGKEVRV PATALCVFDA HDGEVNAVQF SPGSRLLATG GMDRRVKLWE VFGEKCEFKG SLSGSNAGIT SIEFDSAGSY LLAASNDFAS RIWTVDDYRL RHTLTGHSGK VLSAKFLLDN ARIVSGSHDR TLKLWDLRSK VCIKTVFAGS SCNDIVCTEQ CVMSGHFDKK IRFWDIRSES IVREMELLGK ITALDLNPER TELLSCSRDD LLKVIDLRTN AIKQTFSAPG FKCGSDWTRV VFSPDGSYVA AGSAEGSLYI WSVLTGKVEK VLSKQHSSSI NAVAWSPSGS HVVSVDKGCK AVLWAQY | |
Greater than 90% as determined by SDS-PAGE. |