missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_000897.3 |
---|---|
Gene ID (Entrez) | 4881 |
Name | Atrial natriuretic peptide receptor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013260
|
enQuireBio™
QP6431-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16033260
|
enQuireBio™
QP6431-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16023260
|
enQuireBio™
QP6431-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043260
|
enQuireBio™
QP6431-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053260
|
enQuireBio™
QP6431-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16063260
|
enQuireBio™
QP6431-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Spezifikation
NP_000897.3 | |
Atrial natriuretic peptide receptor 1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4881 | |
Research Use Only | |
NPR1 | |
Recombinant Protein | |
E. coli | |
GNLTVAVVLP LANTSYPWSW ARVGPAVELA LAQVKARPDL LPGWTVRTVL GSSENALGVC SDTAAPLAAV DLKWEHNPAV FLGPGCVYAA APVGRFTAHW RVPLLTAGAP ALGFGVKDEY ALTTRAGPSY AKLGDFVAAL HRRLGWERQA LMLYAYRPGD EEHCFFLVEG LFMRVRDRLN ITVDHLEFAE DDLSHYTRLL RTMPRKGRVI YICSSPDAFR TLMLLALEAG LCGEDYVFFH LDIFGQSLQG GQGPAPRRPW ERGDGQDVSA RQAFQAAKII TYKDPDNPEY LEFLKQLKHL AYEQFNFTME DGLVNTIPAS FHDGLLLYIQ AVTETLAHGG TVTDGENITQ RMWNRSFQGV TGYLKIDSSG DRETDFSLWD MDPENGAFRV VLNYNGTSQE LVAVSGRKLN WPLGYPPPDI PKCGFDNEDP ACNQDHLSTL E | |
Greater than 90% as determined by SDS-PAGE. |