missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Annexin A3 Protein

A DNA sequence encoding the Homo sapiens (Human) Annexin A3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8890-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017011

  • 530.00€ / 100µg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_005130.1
Annexin A3 Protein
Research Use Only
ANXA3
Recombinant Protein
E. coli
WVGHRGTVRD YPDFSPSVDA EAIQKAIRGI GTDEKMLISI LTERSNAQRQ LIVKEYQAAY GKELKDDLKG DLSGHFEHLM VALVTPPAVF DAKQLKKSMK GAGTNEDALI EILTTRTSRQ MKDISQAYYT VYKKSLGDDI SSETSGDFRK ALLTLADGRR DESLKVDEHL AKQDAQILYK AGENRWGTDE DKFTEILCLR SFPQLKLTFD EYRNISQKDI VDSIKGELSG HFEDLLLAIV NCVRNTPAFL AERLHRALKG IGTDEFTLNR IMVSRSEIDL LDIRTEFKKH YGYSLYSAIK SDTSGDYEIT LLKICGGD
Greater than 90% as determined by SDS-PAGE.
306
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers