missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Angiopoietin 1 / ANG1 / ANGPT1 Protein
A DNA sequence encoding the Homo sapiens (Human) Angiopoietin 1 / ANG1 / ANGPT1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7600-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001137.2 | |
Angiopoietin 1 / ANG1 / ANGPT1 Protein | |
Research Use Only | |
Angpt1 | |
Recombinant Protein | |
E. coli | |
HIGCSNQRRS PENSGRRYNR IQHGQCAYTF ILPEHDGNCR ESTTDQYNTN ALQRDAPHVE PDFSSQKLQH LEHVMENYTQ WLQKLENYIV ENMKSEMAQI QQNAVQNHTA TMLEIGTSLL SQTAEQTRKL TDVETQVLNQ TSRLEIQLLE NSLSTYKLEK QLLQQTNEIL KIHEKNSLLE HKILEMEGKH KEELDTLKEE KENLQGLVTR QTYIIQELEK QLNRATTNNS VLQKQQLELM DTVHNLVNLC TKEGVLLKGG KREEEKPFRD CADVYQAGFN KSGIYTIYIN NMPEPKKVFC NMDVNGGGWT VIQHREDGSL DFQRGWKEYK MGFGNPSGEY WLGNEFIFAI TSQRQYMLRI ELMDWEGNRA YSQYDRFHIG NEKQNYRLYL KGHTGTAGKQ SSLILHGADF STKDADNDNC MCKCALMLTG GWWFDACGPS NLNGMFYTAG QNHGKLNGIK WHYFKGPSYS LRSTTMMIRP LDF | |
Greater than 90% as determined by SDS-PAGE. |
284 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |