missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Androgen-induced gene 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Androgen-induced gene 1 protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8753-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026301

  • 2385.00€ / 1mg
Estimated Shipment: 28-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001273516.1
Androgen-induced gene 1 Protein
Research Use Only
AIG1
Recombinant Protein
E. coli
TDLSSLLTRG SGNQEQERQL KKLISLRD
Greater than 90% as determined by SDS-PAGE.
51390
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers