missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Anamorsin Protein

A DNA sequence encoding the Homo sapiens (Human) Anamorsin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7733-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020501

Quantity Price
1 2248.90€ / 1mg
3 2115.77€ / 1mg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001295276.1
Anamorsin Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
57019
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CIAPIN1
Recombinant Protein
E. coli
MADFGISAGQ FVAVVWDKSS PVEALKGLVD KLQALTGNEG RVSVENIKQL LQSAHKESSF DIILSGLVPG STTLHSAEIL AEIARILRPG GCLFLKEPVE TAVDNNSKVK TASKLCSALT LSGLVEVKEL QREPLTPEEV QSVREHLGHE SDNLLFVQIT GKKPNFEVGS SRQLKLSITK KSSPSVKPAV DPAAAKLWTL SANDMEDDSM DLIDSDELLD PEDLKKPDPA SLRAASCGEG KKRKACKNCT CGLAEELEKE KSREQMSSQP KSACGNCYLG DAFRCASCPY LGMPAFKPGE KVLLSDSNLH DA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers