missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alpha-tocopherol transfer Protein

A DNA sequence encoding the Homo sapiens (Human) Alpha-tocopherol transfer protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6847-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026260

Quantity Price
1 3211.16€ / 1mg
3 3021.38€ / 1mg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_000361.1
Alpha-tocopherol transfer Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7274
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TTPA
Recombinant Protein
E. coli
MAEARSQPSA GPQLNALPDH SPLLQPGLAA LRRRAREAGV PLAPLPLTDS FLLRFLRARD FDLDLAWRLL KNYYKWRAEC PEISADLHPR SIIGLLKAGY HGVLRSRDPT GSKVLIYRIA HWDPKVFTAY DVFRVSLITS ELIVQEVETQ RNGIKAIFDL EGWQFSHAFQ ITPSVAKKIA AVLTDSFPLK VRGIHLINEP VIFHAVFSMI KPFLTEKIKE RIHMHGNNYK QSLLQHFPDI LPLEYGGEEF SMEDICQEWT NFIMKSEDYL SSISESIQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers