missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Acidic mammalian chitinase Protein

A DNA sequence encoding the Homo sapiens (Human) Acidic mammalian chitinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8133-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042831

Quantity Price
1 493.03€ / 100µg
6 463.44€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001035713.1
Acidic mammalian chitinase Protein
100 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
27159
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CHIA
Recombinant Protein
E. coli
MVSTPENRQT FITSVIKFLR QYEFDGLDFD WEYPGSRGSP PQDKHLFTVL VQEMREAFEQ EAKQINKPRL MVTAAVAAGI SNIQSGYEIP QLSQYLDYIH VMTYDLHGSW EGYTGENSPL YKYPTDTGSN AYLNVDYVMN YWKDNGAPAE KLIVGFPTYG HNFILSNPSN TGIGAPTSGA GPAGPYAKES GIWAYYEICT FLKNGATQGW DAPQEVPYAY QGNVWVGYDN IKSFDIKAQW LKHNKFGGAM VWAIDLDDFT GTFCNQGKFP LISTLKKALG LQSASCTAPA QPIEPITAAP SGSGNGSGSS SSGGSSGGSG FCAVRANGLY PVANNRNAFW HCVNGVTYQQ NCQAGLVFDT SCDCCNWA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers