missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S10 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8417-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024501

  • 530.00€ / 100µg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001005.1
40S ribosomal protein S10 Protein
Research Use Only
RPS10
Recombinant Protein
E. coli
PKKNRIAIYE LLFKEGVMVA KKDVHMPKHP ELADKNVPNL HVMKAMQSLK SRGYVKEQFA WRHFYWYLTN EGIQYLRDYL HLPPEIVPAT LRRSRPETGR PRPKGLEGER PARLTRGEAD RDTYRRSAVP PGADKKAEAG AGSATEFQFR GGFGRGRGQP PQ
Greater than 90% as determined by SDS-PAGE.
6204
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers