missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 14-3-3 tau / 14-3-3 theta / YWHAQ Protein

A DNA sequence encoding the Homo sapiens (Human) 14-3-3 tau / 14-3-3 theta / YWHAQ, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP873-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036181

  • 970.00€ / 200µg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006817.1
14-3-3 tau / 14-3-3 theta / YWHAQ Protein
Research Use Only
YWHAQ
Recombinant Protein
E. coli
MEKTELIQKA KLAEQAERYD DMATCMKAVT EQGAELSNEE RNLLSVAYKN VVGGRRSAWR VISSIEQKTD TSDKKLQLIK DYREKVESEL RSICTTVLEL LDKYLIANAT NPESKVFYLK MKGDYFRYLA EVACGDDRKQ TIDNSQGAYQ EAFDISKKEM QPTHPIRLGL ALNFSVFYYE ILNNPELACT LAKTAFDEAI AELDTLNEDS YKDSTLIMQL LRDNLTLWTS DSAGEECDAA EGAEN
Greater than 80% as determined by SDS-PAGE.
10971
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers