missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 14-3-3 sigma / Stratifin / YWHAS Protein

A DNA sequence encoding the Homo sapiens (Human) 14-3-3 sigma / Stratifin / YWHAS, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP872-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016121

  • 238.00€ / 10µg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006133.1
14-3-3 sigma / Stratifin / YWHAS Protein
Research Use Only
SFN
Recombinant Protein
E. coli
MERASLIQKA KLAEQAERYE DMAAFMKGAV EKGEELSCEE RNLLSVAYKN VVGGQRAAWR VLSSIEQKSN EEGSEEKGPE VREYREKVET ELQGVCDTVL GLLDSHLIKE AGDAESRVFY LKMKGDYYRY LAEVATGDDK KRIIDSARSA YQEAMDISKK EMPPTNPIRL GLALNFSVFH YEIANSPEEA ISLAKTTFDE AMADLHTLSE DSYKDSTLIM QLLRDNLTLW TADNAGEEGG EAPQEPQS
Greater than 90% as determined by SDS-PAGE.
2810
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers