missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 14-3-3 beta / YWHAB Protein
A DNA sequence encoding the Homo sapiens (Human) 14-3-3 beta / YWHAB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP867-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003395.1 | |
14-3-3 beta / YWHAB Protein | |
Research Use Only | |
YWHAB | |
Recombinant Protein | |
E. coli | |
MTMDKSELVQ KAKLAEQAER YDDMAAAMKA VTEQGHELSN EERNLLSVAY KNVVGARRSS WRVISSIEQK TERNEKKQQM GKEYREKIEA ELQDICNDVL ELLDKYLIPN ATQPESKVFY LKMKGDYFRY LSEVASGDNK QTTVSNSQQA YQEAFEISKK EMQPTHPIRL GLALNFSVFY YEILNSPEKA CSLAKTAFDE AIAELDTLNE ESYKDSTLIM QLLRDNLTLW TSENQGDEGD AGEGEN | |
Greater than 90% as determined by SDS-PAGE. |
7529 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |