missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Trans-activator protein BZLF1 Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Trans-activator protein BZLF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7277-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
YP_401673.1 | |
Trans-activator protein BZLF1 Protein | |
Research Use Only | |
BZLF1 | |
Recombinant Protein | |
E. coli | |
MMDPNSTSED VKFTPDPYQV PFVQAFDQAT RVYQDLGGPS QAPLPCVLWP VLPEPLPQGQ LTAYHVSTAP TGSWFSAPQP APENAYQAYA APQLFPVSDI TQNQQTNQAG GEAPQPGDNS TVQTAAAVVF ACPGANQGQQ LADIGVPQPA PVAAPARRTR KPQQPESLEE CDSELEIKRY KNRVASRKCR AKFKQLLQHY REVAAAKSSE NDRLRLLLKQ MCPSLDVDSI IPRTPDVLHE DLLNF | |
Greater than 90% as determined by SDS-PAGE. |
3783744 | |
1 mg | |
Not determined. | |
Full Length | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |