missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Small capsomere-interacting Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Small capsomere-interacting protein, was expressed in the hosts and tags indicated.
440.00€ - 3745.00€
Specifications
Accession Number | YP_401651.1 |
---|---|
Gene ID (Entrez) | 3783701 |
Name | Small capsomere-interacting Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027250
|
enQuireBio™
QP7057-10UG |
10 μg |
440.00€
10µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16067250
|
enQuireBio™
QP7057-50UG |
50 μg |
630.00€
50µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16017250
|
enQuireBio™
QP7057-100UG |
100 μg |
1070.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16047250
|
enQuireBio™
QP7057-200UG |
200 μg |
1765.00€
200µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16057250
|
enQuireBio™
QP7057-500UG |
500 μg |
2360.00€
500µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16037250
|
enQuireBio™
QP7057-1MG |
1 mg |
3745.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Specifications
YP_401651.1 | |
Small capsomere-interacting Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783701 | |
Research Use Only | |
BFRF3 | |
Recombinant Protein | |
E. coli | |
NQNNLPNDVF REAQRSYLVF LTSQFCYEEY VQRTFGVPRR QRAIDKRQRA SVAGAGAHAH LGGSSATPVQ QAQAAASAGT GALASSAPST AVAQSATPSV SSSISSLRAA TSGATAAASA AAAVDTGSGG GGQPHDTAPR GARKKQ | |
Greater than 90% as determined by SDS-PAGE. |