missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7507-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029070

  • 3405.00€ / 1mg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_417389.1
Ribose-5-phosphate isomerase A Protein
Research Use Only
rpiA
Recombinant Protein
E. coli
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK
Greater than 90% as determined by SDS-PAGE.
947407
1 mg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers