missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Probable diguanylate cyclase YedQ Protein
A DNA sequence encoding the Escherichia coli (strain K12) Probable diguanylate cyclase YedQ, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_416465.2 |
---|---|
Gene ID (Entrez) | 946471 |
Name | Probable diguanylate cyclase YedQ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035541
|
enQuireBio™
QP8608-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16055541
|
enQuireBio™
QP8608-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16045541
|
enQuireBio™
QP8608-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065541
|
enQuireBio™
QP8608-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075541
|
enQuireBio™
QP8608-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16085541
|
enQuireBio™
QP8608-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_416465.2 | |
Probable diguanylate cyclase YedQ Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |
946471 | |
Research Use Only | |
yedQ | |
Recombinant Protein | |
E. coli | |
RRMVSNMYVL QSSLQWQAWH DTLTRLYNRG ALFEKARPLA KLCQTHQHPF SVIQVDLDHF KAINDRFGHQ AGDRVLSHAA GLISSSLRAQ DVAGRVGGEE FCVILPGASL TEAAEVAERI RLKLNEKEML IAKSTTIRIS ASLGVSSSEE TGDYDFEQLQ SLADRRLYLA KQAGRNRVFA SDNA | |
Greater than 90% as determined by SDS-PAGE. |