missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Phosphoserine phosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Phosphoserine phosphatase, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_418805.1 |
---|---|
Gene ID (Entrez) | 948913 |
Name | Phosphoserine phosphatase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094070
|
enQuireBio™
QP6548-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16014080
|
enQuireBio™
QP6548-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16004080
|
enQuireBio™
QP6548-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16024080
|
enQuireBio™
QP6548-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034080
|
enQuireBio™
QP6548-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044080
|
enQuireBio™
QP6548-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_418805.1 | |
Phosphoserine phosphatase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
948913 | |
Research Use Only | |
serB | |
Recombinant Protein | |
E. coli | |
MPNITWCDLP EDVSLWPGLP LSLSGDEVMP LDYHAGRSGW LLYGRGLDKQ RLTQYQSKLG AAMVIVAAWC VEDYQVIRLA GSLTARATRL AHEAQLDVAP LGKIPHLRTP GLLVMDMDST AIQIECIDEI AKLAGTGEMV AEVTERAMRG ELDFTASLRS RVATLKGADA NILQQVRENL PLMPGLTQLV LKLETLGWKV AIASGGFTFF AEYLRDKLRL TAVVANELEI MDGKFTGNVI GDIVDAQYKA KTLTRLAQEY EIPLAQTVAI GDGANDLPMI KAAGLGIAYH AKPKVNEKAE VTIRHADLMG VFCILSGSLN QK | |
Greater than 90% as determined by SDS-PAGE. |