missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli nrdB Protein

A DNA sequence encoding the Escherichia coli (strain K12) nrdB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6953-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026860

Quantity Price
1 920.10€ / 100µg
5 865.45€ / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_416738.1
nrdB Protein
100 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
946732
Greater than 90% as determined by SDS-PAGE.
Research Use Only
nrdB
Recombinant Protein
E. coli
AYTTFSQTKN DQLKEPMFFG QPVNVARYDQ QKYDIFEKLI EKQLSFFWRP EEVDVSRDRI DYQALPEHEK HIFISNLKYQ TLLDSIQGRS PNVALLPLIS IPELETWVET WAFSETIHSR SYTHIIRNIV NDPSVVFDDI VTNEQIQKRA EGISSYYDEL IEMTSYWHLL GEGTHTVNGK TVTVSLRELK KKLYLCLMSV NALEAIRFYV SFACSFAFAE RELMEGNAKI IRLIARDEAL HLTGTQHMLN LLRSGADDPE MAEIAEECKQ ECYDLFVQAA QQEKDWADYL FRDGSMIGLN KDILCQYVEY ITNIRMQAVG LDLPFQTRSN PIPWINTWLV SDNVQVAPQE VEVSSYLVGQ IDSEVDTDDL SNFQL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers