missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein II Protein

A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein II, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7296-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018060

  • 3211.16€ / 1mg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_416400.1
Methyl-accepting chemotaxis protein II Protein
1 mg
Not determined.
Cytoplasmic Domain
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
946399
Greater than 90% as determined by SDS-PAGE.
Research Use Only
tar
Recombinant Protein
E. coli
IRRMLLTPLA KIIAHIREIA GGNLANTLTI DGRSEMGDLA QSVSHMQRSL TDTVTHVREG SDAIYAGTRE IAAGNTDLSS RTEQQASALE ETAASMEQLT ATVKQNADNA RQASQLAQSA SDTAQHGGKV VDGVVKTMHE IADSSKKIAD IISVIDGIAF QTNILALNAA VEAARAGEQG RGFAVVAGEV RNLASRSAQA AKEIKALIED SVSRVDTGSV LVESAGETMN NIVNAVTRVT DIMGEIASAS DEQSRGIDQV ALAVSEMDRV TQQNASLVQE SAAAAAALEE QASRLTQAVS AFRLAASPLT NKPQTPSRPA SEQPPAQPRL RIAEQDPNWE TF
Videos
SDS
Documents
Certificates
Special Offers

Special Offers