missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli L-lactate dehydrogenase Protein
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) L-lactate dehydrogenase, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | WP_000586962.1 |
---|---|
Name | L-lactate dehydrogenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | lldD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008860
|
enQuireBio™
QP7434-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16028860
|
enQuireBio™
QP7434-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16018860
|
enQuireBio™
QP7434-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038860
|
enQuireBio™
QP7434-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048860
|
enQuireBio™
QP7434-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058860
|
enQuireBio™
QP7434-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Spezifikation
WP_000586962.1 | |
Research Use Only | |
lldD | |
Recombinant Protein | |
E. coli | |
MIISAASDYR AAAQRILPPF LFHYMDGGAY SEYTLRRNVE DLSEVALRQR ILKNMSDLSL ETTLFNEKLS MPVALAPVGL CGMYARRGEV QAAKAADAHG IPFTLSTVSV CPIEEVAPAI KRPMWFQLYV LRDRGFMRNA LERAKAAGCS TLVFTVDMPT PGARYRDAHS GMSGPNAAMR RYLQAVTHPQ WAWDVGLNGR PHDLGNISAY LGKPTGLEDY IGWLGNNFDP SISWKDLEWI RDFWDGPMVI KGILDPEDAR DAVRFGADGI VVSNHGGRQL DGVLSSARAL PAIADAVKGD IAILADSGIR NGLDVVRMIA LGADTVLLGR AFLYALATAG QAGVANLLNL IEKEMKVAMT LTGAKSISEI TQDSLVQGLG KELPAALAPM AKGNAA | |
Greater than 90% as determined by SDS-PAGE. |
L-lactate dehydrogenase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |