Learn More
enQuireBio™ Recombinant E. coli Heat-labile enterotoxin B chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Heat-labile enterotoxin B chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7016-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_024167713.1 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Heat-labile enterotoxin B chain Protein | |
Research Use Only | |
eltB | |
Recombinant Protein | |
E. coli | |
APQSITELCS EYHNTQIYTI NDKILSYTES MAGKREMVII TFKSGATFQV EVPGSQHIDS QKKAIERMKD TLRITYLTET KIDKLCVWNN KTPNSIAAIS MEN | |
Greater than 90% as determined by SDS-PAGE. |