missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glucose-6-phosphate 1-dehydrogenase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glucose-6-phosphate 1-dehydrogenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8937-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017241

  • 3211.16€ / 1mg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_416366.1
Glucose-6-phosphate 1-dehydrogenase Protein
1 mg
Not determined.
Partial (see sequence information for more details).
E. coli
GST
Tris-based buffer, 50% glycerol
946370
Greater than 90% as determined by SDS-PAGE.
Research Use Only
zwf
Recombinant Protein
E. coli
MAVTQTAQAC DLVIFGAKGD LARRKLLPSL YQLEKAGQLN PDTRIIGVGR ADWDKAAYTK VVREALETFM KETIDEGLWD TLSARLDFCN LDVNDTAAFS RLGAMLDQKN RITINYFAMP PSTFGAICKG LGEAKLNAKP ARVVMEKPLG TSLATSQEIN DQVGEYFEEC QVYRIDHYLG KETVLNLLAL RFANSLFVNN WDNRTIDHVE ITVAEEVGIE GRWGYFDKAG QMRDMIQNHL LQILCMIAMS PPSDLSADSI RDEKVKVLKS LRRIDRSNVR EKTVRGQYTA GFAQGKKVPG YLEEEGANKS SNTETFVAIR VDIDNWRWAG VPFYLRTGKR LPTKCSEVVV YFKTPELNLF KESWQDLPQN KLTIRLQPDE GVDIQVLNKV PGLDHKHNLQ ITKLDLSYSE TFNQTHLADA YERLLLETMR GIQALFVRRD EVEEAWKWVD SITEAWAMDN DAPKPYQAGT WGPVASVAMI TRDGRS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers