missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Beta-lactamase TEM Protein

A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase TEM, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7256-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037890

  • 3849.07€ / 1mg

Estimated Shipment: 18-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_943295.1
Beta-lactamase TEM Protein
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
10076131
Greater than 90% as determined by SDS-PAGE.
Research Use Only
bla
Recombinant Protein
Yeast
HPETLVKVKD AEDQLGARVG YIELDLNSGK ILESFRPEER FPMMSTFKVL LCGAVLSRVD AGQEQLGRRI HYSQNDLVEY SPVTEKHLTD GMTVRELCSA AITMSDNTAA NLLLTTIGGP KELTAFLHNM GDHVTRLDRW EPELNEAIPN DERDTTMPAA MATTLRKLLT GELLTLASRQ QLIDWMEADK VAGPLLRSAL PAGWFIADKS GAGERGSRGI IAALGPDGKP SRIVVIYTTG SQATMDERNR QIAEIGASLI KHW
Videos
SDS
Documents
Certificates
Special Offers

Special Offers