missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Beta-lactamase TEM Protein
A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase TEM, was expressed in the hosts and tags indicated.
352.00€ - 4075.00€
Specifications
Accession Number | NP_943295.1 |
---|---|
Gene ID (Entrez) | 10076131 |
Name | Beta-lactamase TEM Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017890
|
enQuireBio™
QP7256-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16037890
|
enQuireBio™
QP7256-YE-1MG |
1 mg |
4075.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16027890
|
enQuireBio™
QP7256-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047890
|
enQuireBio™
QP7256-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16057890
|
enQuireBio™
QP7256-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067890
|
enQuireBio™
QP7256-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_943295.1 | |
Beta-lactamase TEM Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
10076131 | |
Research Use Only | |
bla | |
Recombinant Protein | |
Yeast | |
HPETLVKVKD AEDQLGARVG YIELDLNSGK ILESFRPEER FPMMSTFKVL LCGAVLSRVD AGQEQLGRRI HYSQNDLVEY SPVTEKHLTD GMTVRELCSA AITMSDNTAA NLLLTTIGGP KELTAFLHNM GDHVTRLDRW EPELNEAIPN DERDTTMPAA MATTLRKLLT GELLTLASRQ QLIDWMEADK VAGPLLRSAL PAGWFIADKS GAGERGSRGI IAALGPDGKP SRIVVIYTTG SQATMDERNR QIAEIGASLI KHW | |
Greater than 90% as determined by SDS-PAGE. |