missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7047-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027190

  • 1037.95€ / 100µg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_417884.1
Aerobic glycerol-3-phosphate dehydrogenase Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
947934
Greater than 90% as determined by SDS-PAGE.
Research Use Only
glpD
Recombinant Protein
Yeast
METKDLIVIG GGINGAGIAA DAAGRGLSVL MLEAQDLACA TSSASSKLIH GGLRYLEHYE FRLVSEALAE REVLLKMAPH IAFPMRFRLP HRPHLRPAWM IRIGLFMYDH LGKRTSLPGS TGLRFGANSV LKPEIKRGFE YSDCWVDDAR LVLANAQMVV RKGGEVLTRT RATSARRENG LWIVEAEDID TGKKYSWQAR GLVNATGPWV KQFFDDGMHL PSPYGIRLIK GSHIVVPRVH TQKQAYILQN EDKRIVFVIP WMDEFSIIGT TDVEYKGDPK AVKIEESEIN YLLNVYNTHF KKQLSRDDIV WTYSGVRPLC DDESDSPQAI TRDYTLDIHD ENGKAPLLSV FGGKLTTYRK LAEHALEKLT PYYQGIGPAW TKESVLPGGA IEGDRDDYAA RLRRRYPFLT ESLARHYART YGSNSELLLG NAGTVSDLGE DFGHEFYEAE LKYLVDHEWV RRADDALWRR TKQGMWLNAD QQSRVSQWLV EYTQQRLSLA S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers