missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L35 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L35, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8626-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015651

Quantity Price
1 920.10€ / 100µg
5 865.45€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_416232.3
50S ribosomal protein L35 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
E. coli
GST
Tris-based buffer, 50% glycerol
32367223
Greater than 90% as determined by SDS-PAGE.
Research Use Only
rpmI
Recombinant Protein
E. coli
VRGAAKRFKK TGKGGFKHKH ANLRHILTKK ATKRKRHLRP KAMVSKGDLG LVIACLPY
Videos
SDS
Documents
Certificates
Special Offers

Special Offers