missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis omcA Protein
A DNA sequence encoding the Chlamydia trachomatis omcA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7018-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_219956.1 | |
omcA Protein | |
Research Use Only | |
omcA | |
Recombinant Protein | |
E. coli | |
CCRIVDCCFE DPCAPIQCSP CESKKKDVDG GCNSCNGYVP ACKPCGGDTH QDAKHGPQAR GIPVDGKCRQ | |
Greater than 90% as determined by SDS-PAGE. |
884216 | |
100 μg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |