missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Large cysteine-rich periplasmic protein OmcB Protein
A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) Large cysteine-rich periplasmic protein OmcB, was expressed in the hosts and tags indicated.
137.00€ - 2450.00€
Specifications
Accession Number | NP_219955.1 |
---|---|
Gene ID (Entrez) | 884223 |
Name | Large cysteine-rich periplasmic protein OmcB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16017050
|
enQuireBio™
QP7015-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16057050
|
enQuireBio™
QP7015-50UG |
50 μg |
339.00€
50µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16007050
|
enQuireBio™
QP7015-100UG |
100 μg |
545.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16037050
|
enQuireBio™
QP7015-200UG |
200 μg |
870.00€
200µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16047050
|
enQuireBio™
QP7015-500UG |
500 μg |
1500.00€
500µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16027050
|
enQuireBio™
QP7015-1MG |
1 mg |
2450.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Especificaciones
NP_219955.1 | |
Large cysteine-rich periplasmic protein OmcB Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
884223 | |
Research Use Only | |
omcB | |
Recombinant Protein | |
E. coli | |
LADTKAKDNT SHKSKKARKN HSKETPVDRK EVAPVHESKA TGPKQDSCFG RMYTVKVNDD RNVEITQAVP EYATVGSPYP IEITATGKRD CVDVIITQQL PCEAEFVRSD PATTPTADGK LVWKIDRLGQ GEKSKITVWV KPLKEGCCFT AATVCA | |
Greater than 90% as determined by SDS-PAGE. |