missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum clpP Protein

A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) clpP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7429-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028830

  • 3405.00€ / 1mg
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

WP_003357557.1
clpP Protein
Research Use Only
clpP
Recombinant Protein
E. coli
MSLVPVVVEQ TNRGERSYDI YSRLLKDRII MLSEEVNDTT ASLIVAQLLF LEAEDPDKDI HLYINSPGGS ITSGMAIYDT MQYIKPDVST ICVGMAASMG AFLLAAGAKG KRYALPNSEV MIHQPLGGFR GQATDIGIHA ERILKMKKKL NTILSDRTGK PLEQVELDTE RDHFLSAEEA KEYGLIDEVI DKKK
Greater than 90% as determined by SDS-PAGE.
5187372
1 mg
Not determined.
Full Length
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers