missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum clpP Protein
A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) clpP, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | WP_003357557.1 |
---|---|
Gene ID (Entrez) | 5187372 |
Name | clpP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008830
|
enQuireBio™
QP7429-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16028830
|
enQuireBio™
QP7429-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16018830
|
enQuireBio™
QP7429-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038830
|
enQuireBio™
QP7429-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048830
|
enQuireBio™
QP7429-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058830
|
enQuireBio™
QP7429-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
WP_003357557.1 | |
clpP Protein | |
Not determined. | |
Full Length | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5187372 | |
Research Use Only | |
clpP | |
Recombinant Protein | |
E. coli | |
MSLVPVVVEQ TNRGERSYDI YSRLLKDRII MLSEEVNDTT ASLIVAQLLF LEAEDPDKDI HLYINSPGGS ITSGMAIYDT MQYIKPDVST ICVGMAASMG AFLLAAGAKG KRYALPNSEV MIHQPLGGFR GQATDIGIHA ERILKMKKKL NTILSDRTGK PLEQVELDTE RDHFLSAEEA KEYGLIDEVI DKKK | |
Greater than 90% as determined by SDS-PAGE. |