missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type C1 Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type C1, was expressed in the hosts and tags indicated.
440.00€ - 3745.00€
Specifications
Name | Botulinum neurotoxin type C1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Partial (see sequence information for more details). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087390
|
enQuireBio™
QP7104-10UG |
10 μg |
440.00€
10µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16027400
|
enQuireBio™
QP7104-50UG |
50 μg |
630.00€
50µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16077390
|
enQuireBio™
QP7104-100UG |
100 μg |
1070.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16007400
|
enQuireBio™
QP7104-200UG |
200 μg |
1765.00€
200µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16017400
|
enQuireBio™
QP7104-500UG |
500 μg |
2360.00€
500µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16097390
|
enQuireBio™
QP7104-1MG |
1 mg |
3745.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Specifications
Botulinum neurotoxin type C1 Protein | |
Not determined. | |
Recombinant Protein | |
E. coli | |
PITINNFNYS DPVDNKNILY LDTHLNTLAN EPEKAFRITG NIWVIPDRFS RNSNPNLNKP PRVTSPKSGY YDPNYLSTDS DKDPFLKEII KLFKRINSRE IGEELIYRLS TDIPFPGNNN TPINTFDFDV DFNSVDVKTR QGNNWVKTGS INPSVIITGP RENIIDPETS TFKLTNNTFA AQEGFGALSI ISISPRFMLT YSNATNDVGE GRFSKSEFCM DPILILMHEL NHAMHNLYGI AIPNDQTISS VTSNIFYSQY NVKLEYAEIY AFGGPTIDLI PKSARKYFEE KALDYYRSIA KRLNSITTAN PSSFNKYIGE YKQKLIRKYR FVVESSGEVT VNRNKFVELY NELTQIFTEF NYAKIYNVQN RKIYLSNVYT PVTANILDDN VYDIQNGFNI PKSNLNVLFM GQNLSRNPAL RKVNPEN | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Partial (see sequence information for more details). | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |