missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type B Protein

A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9517-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018431

Quantity Price
1 2497.57€ / 500µg
3 2349.71€ / 500µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

Botulinum neurotoxin type B Protein
500 μg
Not determined.
Partial (see sequence information for more details).
C. botulinum
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
botB
Recombinant Protein
Yeast
MPVTINNFNY NDPIDNNNII MMEPPFARGT GRYYKAFKIT DRIWIIPERY TFGYKPEDFN KSSGIFNRDV CEYYDPDYLN TNDKKNIFLQ TMIKLFNRIK SKPLGEKLLE MIINGIPYLG DRRVPLEEFN TNIASVTVNK LISNPGEVER KKGIFANLII FGPGPVLNEN ETIDIGIQNH FASREGFGGI MQMKFCPEYV SVFNNVQENK GASIFNRRGY FSDPALILMH ELIHVLHGLY GIKVDDLPIV PNEKKFFMQS TDAIQAEELY TFGGQDPSII TPSTDKSIYD KVLQNFRGIV DRLNKVLVCI SDPNININIY KNKFKDKYKF VEDSEGKYSI DVESFDKLYK SLMFGFTETN IAENYKIKTR ASYFSDSLPP VKIKNLLDNE IYTIEEGFNI SDKDMEKEYR GQNKAINKQA YEEISKE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers