missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine LH-beta Protein

A DNA sequence encoding the Bos taurus (Bovine) LH-beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6303-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022320

Quantity Price
1 920.10€ / 100µg
5 865.45€ / 100µg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_776355.1
LH-beta Protein
100 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
280839
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LHB
Recombinant Protein
E. coli
SRGPLRPLCQ PINATLAAEK EACPVCITFT TSICAGYCPS MKRVLPVILP PMPQRVCTYH ELRFASVRLP GCPPGVDPMV SFPVALSCHC GPCRLSSTDC GGPRTQPLAC DHPPLPDILF L
Videos
SDS
Documents
Certificates
Special Offers

Special Offers