missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Agouti-signaling Protein

A DNA sequence encoding the Bos taurus (Bovine) Agouti-signaling protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7635-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019930

Quantity Price
1 920.10€ / 100µg
5 865.45€ / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_996674.1
Agouti-signaling Protein
100 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
404192
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ASIP
Recombinant Protein
E. coli
HLAPEEKPRD ERNLKNNSSM NLLDFPSVSI VALNKKSKKI SRNEAEKKKR PSKRKAPMKN VARTRPPPPT PCVATRDSCK PPAPACCDPC AFCQCRFFRS ACSCRVLNPT C
Videos
SDS
Documents
Certificates
Special Offers

Special Offers