missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rat CTLA-4 / CD152 Protein, Fc Tag

326.60€ - 2325.44€

Specifications

Molecular Weight (g/mol) 40.2 kDa
Source Rat CTLA-4, Fc Tag (CT4-R5259) is expressed from human 293 cells (HEK293). It contains AA Ile 38 - Asp 161 (Accession # Q62859).
Immunogen Ile 38 - Asp 161
Storage Requirements -20°C
Gene Alias CTLA4,CD152
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15836537
View Documents Promotion Details
ACROBiosystems
CT4-R5259-1MG
1 mg
2325.44€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
15826537
View Documents Promotion Details
ACROBiosystems
CT4-R5259-100UG
100 μg
326.60€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
 
Description

Description

CTLA-4 (Cytotoxic T-Lymphocyte Antigen 4) is also known as CD152 (Cluster of differentiation 152), is a protein receptor that downregulates the immune system. CTLA4 is a member of the immunoglobulin superfamily, which is expressed on the surface of Helper T cells and transmits an inhibitory signal to T cells. The protein contains an extracellular V domain, a transmembrane domain, and a cytoplasmic tail. Alternate splice variants, encoding different isoforms. CTLA4 is similar to the T-cell co-stimulatory protein, CD28, and both molecules bind to CD80 and CD86, also called B7-1 and B7-2 respectively, on antigen-presenting cells. CTLA4 transmits an inhibitory signal to T cells, whereas CD28 transmits a stimulatory signal. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function. Fusion proteins of CTLA4 and antibodies (CTLA4-Ig) have been used in clinical trials for rheumatoid arthritis.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE).
Specifications

Specifications

40.2 kDa
Ile 38 - Asp 161
CTLA4,CD152
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Rat CTLA-4, Fc Tag (CT4-R5259) is expressed from human 293 cells (HEK293). It contains AA Ile 38 - Asp 161 (Accession # Q62859).
-20°C
CTLA-4
Rat
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins
Videos
SDS
Documents

Documents

Product Certifications