Learn More
Novus Biologicals™ Rat Creatine Kinase MB ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75313
Product Code. 18392580
  • 836.00€ / 1 set
Description
The Rat Creatine Kinase MB ELISA Kit (Colorimetric) measures rat Creatine Kinase MB in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
31.25 to 2000 pg/mL
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed.
Other Biological Fluids, Plasma, Plasma, Serum, Serum
ELISA
CKB
1 Kit
Storage of components varies. See protocol for specific instructions.
Creatine Kinase MB
18.75 pg/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed.
1152
Rat
metabolism
Creatine Kinase Isoenzyme