Learn More
Novus Biologicals™ Rat Creatine Kinase MB ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75313
This item is not returnable.
View return policy
Description
The Rat Creatine Kinase MB ELISA Kit (Colorimetric) measures rat Creatine Kinase MB in serum, plasma and other biological fluids.
Specifications
31.25 to 2000 pg/mL | |
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed. | |
Other Biological Fluids, Plasma, Plasma, Serum, Serum | |
ELISA | |
CKB | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Creatine Kinase MB |
18.75 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat CKMB in samples. No significant cross-reactivity or interference between Rat CKMB and analogues was observed. | |
1152 | |
Rat | |
metabolism | |
Creatine Kinase Isoenzyme |