Learn More
IBA Solutions For Life Science Random RNA-PTO-Hexamer
Pre-made random RNA-oligonucleotides, phosphothioate (PTO) sequences manufactured in state-of-the art facilities for use in cell-free cloning based on the multiple-primed rolling circle amplification (MPRCA) technique.
Brand:  IBA Solutions For Life Science 5-0411-213
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15623618
  • 226.00€ / 200nmol
Estimated Shipment date
from Supplier 29-05-2019
Add to basket
Description
Description
RNA-PTO oligonucleotides avoid the formation of unwanted ligation products that form if DNA primers are used.
  • Due to the phosphorothioate modifications, the stability is increased significantly
  • Superior performance in cell culture assays looking for an immune response
  • Avoids any kind of endotoxin-related unspecific effects
  • Used to identify Toll-like receptors (TLR) as sensors for single stranded RNA
  • Manufactured using the highest quality raw materials, with regular quality checks
Specifications
Specifications
For cell-free cloning based on the MPRCA technique
Random RNA-oligonucleotides, phosphothioate (PTO) protected
200 nmol
Videos