Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27414
Additional Details : Weight : 3.65000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
25 mm | |
Upright Freezers | |
10 x 4 |
360 x 135 x 265 mm | |
Microtiterplates (40) | |
Stainless Steel |