missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53I11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TP53I11.
Brand: Abnova PAB17335.100ug
Additional Details : Weight : 0.02000kg
Specifications
TP53I11 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of TP53I11. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
PIG11 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total TP53I11 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
TP53I11 | |
TP53I11 | |
A synthetic peptide corresponding to internal of human TP53I11. | |
RUO | |
9537 | |
Human | |
Liquid |
For Research Use Only