missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NDRG3.
Brand: Abnova PAB21027.100uL
Additional Details : Weight : 0.02000kg
Description
Sequence: CAKGWIDWAASKLSGLTTNVVDIILAHHFGQEELQANLDLIQTYRMHIAQDINQDNLQLFLNSYNGRRDLEIERPILGQNDNKSKTLKCSTLLVVGDSpecifications
NDRG3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant NDRG3. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
FLJ13556 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
57446 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
NDRG3 | |
NDRG3 | |
Recombinant protein corresponding to amino acids of human NDRG3. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only