Learn More
ADD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADD2.
Brand: Abnova PAB18059.100ug
Additional Details : Weight : 0.02000kg
Description
Adducins are heteromeric proteins composed of different subunits referred to as adducin alpha, beta and gamma. The three subunits are encoded by distinct genes and belong to a family of membrane skeletal proteins involved in the assembly of spectrin-actin network in erythrocytes and at sites of cell-cell contact in epithelial tissues. While adducins alpha and gamma are ubiquitously expressed, the expression of adducin beta is restricted to brain and hematopoietic tissues. Adducin, originally purified from human erythrocytes, was found to be a heterodimer of adducins alpha and beta. Polymorphisms resulting in amino acid substitutions in these two subunits have been associated with the regulation of blood pressure in an animal model of hypertension. Heterodimers consisting of alpha and gamma subunits have also been described. Structurally, each subunit is comprised of two distinct domains. The amino-terminal region is protease resistant and globular in shape, while the carboxy-terminal region is protease sensitive. The latter contains multiple phosphorylation sites for protein kinase C, the binding site for calmodulin, and is required for association with spectrin and actin. Various adducin beta mRNAs, alternatively spliced at 3'end and/or internally spliced and encoding different isoforms, have been described. The functions of all the different isoforms are not known. [provided by RefSeq]
Specifications
ADD2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ADD2. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P35612 | |
ADD2 | |
A synthetic peptide corresponding to human ADD2. | |
RUO | |
119 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Western Blot (1:500∼1:1000) Immunofluorescence (1:500∼1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
ADD2 | |
ADDB | |
Rabbit | |
100 μg | |
Primary | |
This antibody is specific to ADD2. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only