missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-N-Acetyl-α-allylglycine, 98%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 101°C to 106°C |
Color | White to Yellow |
Enantiomeric Excess | 98.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Quantity | 100 mg |
Specific Rotation | -8.3 |
Formula Weight | 157.17 |
Show More |
Safety and Handling
Product Identifier
- (R)-N-Acetyl-alpha-allylglycine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'